Thermo Scientific Chemicals

trans-4-Cyanocyclohexylamine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: H62522.03
1 g, Each
Thermo Scientific Chemicals

trans-4-Cyanocyclohexylamine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: H62522.03
1 g, Each
Quantity
Catalog number: H62522.03
also known as H62522-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
873537-33-8
IUPAC Name
4-aminocyclohexane-1-carbonitrile hydrochloride
Molecular Formula
C7H13ClN2
InChI Key
BTLVSLNZSMIPCW-UHFFFAOYSA-N
SMILES
Cl.NC1CCC(CC1)C#N
Appearance (Color)
White
Assay (GC)
>97%
Proton NMR
Conforms to structure
Carbon-13 NMR
Conforms to structure
Form
Solid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Notes
Store in a cool and dry condition. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text