Thermo Scientific Chemicals

N-Boc-1-Boc-L-tryptophan, 95%, Thermo Scientific Chemicals

Catalog number: H62542.03
1 g, Each
Thermo Scientific Chemicals

N-Boc-1-Boc-L-tryptophan, 95%, Thermo Scientific Chemicals

Catalog number: H62542.03
1 g, Each
Quantity
Catalog number: H62542.03
also known as H62542-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
144599-95-1
IUPAC Name
(2S)-3-{1-[(tert-butoxy)carbonyl]-1H-indol-3-yl}-2-{[(tert-butoxy)carbonyl]amino}propanoic acid
Molecular Formula
C21H28N2O6
InChI Key
FATGZMFSCKUQGO-HNNXBMFYSA-N
SMILES
CC(C)(C)OC(=O)N[C@@H](CC1=CN(C(=O)OC(C)(C)C)C2=CC=CC=C12)C(O)=O
Appearance (Color)
White to pale cream
Form
Powder
Assay from Supplier's CofA
≥94.0% (HPLC)
Identification (FTIR)
Conforms
Melting Point (clear melt)
158-160°C

Description

N-Boc-1-Boc-L-tryptophan is generally used as a intermediate in pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-1-Boc-L-tryptophan is generally used as a intermediate in pharmaceutical and chemical research.

Solubility
Soluble in dimethylformamide (1 mmole in 2 ml).

Notes
Store at -20°C. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text