Thermo Scientific Chemicals

N-Boc-L-aspartic acid 1-methyl ester, 98%, Thermo Scientific Chemicals

Catalog number: H62590.03
1 g, Each
Thermo Scientific Chemicals

N-Boc-L-aspartic acid 1-methyl ester, 98%, Thermo Scientific Chemicals

Catalog number: H62590.03
1 g, Each
Quantity
Catalog number: H62590.03
also known as H62590-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
98045-03-5
Assay (HPLC)
>97.5%

Description

N-Boc-L-aspartic acid 1-methyl ester is used as an intermediate in the preparation of modified alfa-amino acids and 5-cis-alkylprolines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-L-aspartic acid 1-methyl ester is used as an intermediate in the preparation of modified alfa-amino acids and 5-cis-alkylprolines.

Notes
Incompatible with oxidizing agents. Heat sensitive.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text