N-Boc-L-aspartic acid 1-methyl ester is used as an intermediate in the preparation of modified alfa-amino acids and 5-cis-alkylprolines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Boc-L-aspartic acid 1-methyl ester is used as an intermediate in the preparation of modified alfa-amino acids and 5-cis-alkylprolines.
Notes
Incompatible with oxidizing agents. Heat sensitive.
RUO – Research Use Only
Mohite, A. R.; Bhat, R. G. Enantiopure Synthesis of Side Chain-Modified alpha-Amino Acids and 5-cis-Alkylprolines. J. Org. Chem. 2012, 77 (12), 5423-5428.