Thermo Scientific Chemicals

N-Boc-L-alaninamide, 96%, Thermo Scientific Chemicals

Catalog number: H62604.06
5 g, Each
Thermo Scientific Chemicals

N-Boc-L-alaninamide, 96%, Thermo Scientific Chemicals

Catalog number: H62604.06
5 g, Each
Quantity
Catalog number: H62604.06
also known as H62604-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
85642-13-3
IUPAC Name
tert-butyl N-(1-carbamoylethyl)carbamate
Molecular Formula
C8H16N2O3
InChI Key
GZKKSKKIABUUCX-UHFFFAOYNA-N
SMILES
CC(NC(=O)OC(C)(C)C)C(N)=O
Assay (HPLC)
>95.0%

Description

N-Boc-L-alaninamide is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-L-alaninamide is used as pharmaceutical intermediate.

Solubility
Soluble in water or 1% acetic acid.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text