N-Boc-glycine ethyl ester, 95%, Thermo Scientific Chemicals
N-Boc-glycine ethyl ester, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-glycine ethyl ester, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H62611.06
also known as H62611-06
Price (USD)/ Each
63.40
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
63.40
Add to cart
N-Boc-glycine ethyl ester, 95%, Thermo Scientific Chemicals
Catalog numberH62611.06
Price (USD)/ Each
63.40
-
Add to cart
Chemical Identifiers
CAS14719-37-0
IUPAC Nameethyl 2-{[(tert-butoxy)carbonyl]amino}acetate
Molecular FormulaC9H17NO4
InChI KeyCNIBHMMDDXGDNR-UHFFFAOYSA-N
SMILESCCOC(=O)CNC(=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only