D-Cysteine is an important source of sulfide in human metabolism. It is the precursor for glutathione. It plays important role in protein structure and in metal binding.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
D-Cysteine is an important source of sulfide in human metabolism. It is the precursor for glutathione. It plays important role in protein structure and in metal binding.
Solubility
Soluble in water (partly).
Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.
RUO – Research Use Only
Klaas R. Krijgsheld; Eltjo J. Glazenburg; Egbert Scholtens; Gerard J. Mulder. The oxidation of L- and D-cysteine to inorganic sulfate and taurine in the rat. Biochimica et Biophysica Acta (BBA) - General Subjects. 1981, 677 (1), (13), 7-12.
Kenji Yamaguchi; Shigeki Sakakibara; Juro Asamizu; Iwao Ueda. Induction and activation of cysteine oxidase of rat liver II. The measurement of cysteine metabolism in vivo and the activation of in vivo activity of cysteine oxidase. Biochimica et Biophysica Acta (BBA) - General Subjects. 1973, 297 (1), 48-59.