D-Cysteine, 98%, Thermo Scientific Chemicals
D-Cysteine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

D-Cysteine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number H62627.06
also known as H62627-06
Price (USD)/ Each
325.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
325.00
Add to cart
D-Cysteine, 98%, Thermo Scientific Chemicals
Catalog numberH62627.06
Price (USD)/ Each
325.00
-
Add to cart
Chemical Identifiers
CAS921-01-7
IUPAC Name(2S)-2-amino-3-sulfanylpropanoic acid
Molecular FormulaC3H7NO2S
InChI KeyXUJNEKJLAYXESH-RUOZJJLTNA-N
SMILESN[C@H](CS)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder
Assay (Titration)>97.5%
Appearance (Color)White to off-white
D-Cysteine is an important source of sulfide in human metabolism. It is the precursor for glutathione. It plays important role in protein structure and in metal binding.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Cysteine is an important source of sulfide in human metabolism. It is the precursor for glutathione. It plays important role in protein structure and in metal binding.

Solubility
Soluble in water (partly).

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.
RUO – Research Use Only

General References:

  1. Klaas R. Krijgsheld; Eltjo J. Glazenburg; Egbert Scholtens; Gerard J. Mulder. The oxidation of L- and D-cysteine to inorganic sulfate and taurine in the rat. Biochimica et Biophysica Acta (BBA) - General Subjects. 1981, 677 (1), (13), 7-12.
  2. Kenji Yamaguchi; Shigeki Sakakibara; Juro Asamizu; Iwao Ueda. Induction and activation of cysteine oxidase of rat liver II. The measurement of cysteine metabolism in vivo and the activation of in vivo activity of cysteine oxidase. Biochimica et Biophysica Acta (BBA) - General Subjects. 1973, 297 (1), 48-59.