Thermo Scientific Chemicals

N-Fmoc-D-aspartic acid 1-tert-butyl ester, 95%, Thermo Scientific Chemicals

Catalog number: H62680.06
5 g, Each
Thermo Scientific Chemicals

N-Fmoc-D-aspartic acid 1-tert-butyl ester, 95%, Thermo Scientific Chemicals

Catalog number: H62680.06
5 g, Each
Quantity
Catalog number: H62680.06
also known as H62680-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
134098-70-7
IUPAC Name
4-(tert-butoxy)-3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-oxobutanoic acid
Molecular Formula
C23H25NO6
InChI Key
VZXQYACYLGRQJU-UHFFFAOYNA-N
SMILES
CC(C)(C)OC(=O)C(CC(O)=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Assay (HPLC)
>94.0%

Description

N-Fmoc-D-aspartic acid 1-tert-butyl ester is an pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-D-aspartic acid 1-tert-butyl ester is an pharmaceutical intermediate.

Solubility
Soluble in DMF.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text