N-Boc-O-tert-butyl-L-threonine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Boc-O-tert-butyl-L-threonine is used as pharmaceutical intermediate.
Solubility
Soluble in DMF.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only
Antonio Procopio; Renato Dalpozzo; Antonio De Nino; Loredana Maiuolo; Monica Nardi; Manuela Oliverio; Beatrice Russo. A facile Er(OTf)3-catalyzed synthesis of 2,3-unsaturated O- and S-glycosides. Carbohydrate Research. 2007, 342 (14), 2125-2131.