N-Boc-D-glutamic acid, 98%, Thermo Scientific Chemicals
N-Boc-D-glutamic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-D-glutamic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H62762.06
also known as H62762-06
Price (USD)/ Each
80.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
94.90 
Save 14.25 (15%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
80.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
94.90 
Save 14.25 (15%)
Add to cart
N-Boc-D-glutamic acid, 98%, Thermo Scientific Chemicals
Catalog numberH62762.06
Price (USD)/ Each
80.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
94.90 
Save 14.25 (15%)
-
Add to cart
Chemical Identifiers
CAS34404-28-9
IUPAC Name2-{[(tert-butoxy)carbonyl]amino}pentanedioic acid
Molecular FormulaC10H17NO6
InChI KeyAQTUACKQXJNHFQ-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)NC(CCC(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormSolid
Appearance (Color)White
Assay (unspecified)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only