N-Fmoc-O-tert-butyl-D-tyrosine is potentially useful for proteomics studies and solid phase peptide synthesis techniques.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Fmoc-O-tert-butyl-D-tyrosine is potentially useful for proteomics studies and solid phase peptide synthesis techniques.
Solubility
Soluble in 1 mmole in 2 ml dimethylformamide (Clearly soluble). Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only
Mario Gonçalves; Karine Estieu-Gionnet; Georges Laïn; Mireille Bayle; Natacha Betz; Gérard Déléris. On-resin cyclization of a head-to-tail cyclopeptide using an allyldimethylsilyl polystyrene resin pre-loaded by metathesis. Tetrahedron. 2005, 61 (32), 7789-7795.
Holger Kubas; Martin Schäfer; Ulrike Bauder-Wüst; Matthias Edera; Dörte Oltmanns; Uwe Haberkorn; Walter Mier; Michael Eisenhuts. Multivalent cyclic RGD ligands: influence of linker lengths on receptor binding. Nuclear Medicine and Biology. 2005, 8 (37), 885-891.