Nalpha,Nepsilon-Di-Boc-L-lysine dicyclohexylammonium salt, 98%, Thermo Scientific Chemicals
Nalpha,Nepsilon-Di-Boc-L-lysine dicyclohexylammonium salt, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nalpha,Nepsilon-Di-Boc-L-lysine dicyclohexylammonium salt, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number H62785.22
also known as H62785-22
Price (USD)/ Each
152.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
152.00
Add to cart
Nalpha,Nepsilon-Di-Boc-L-lysine dicyclohexylammonium salt, 98%, Thermo Scientific Chemicals
Catalog numberH62785.22
Price (USD)/ Each
152.00
-
Add to cart
Chemical Identifiers
CAS15098-69-8
IUPAC Name(2S)-2,6-bis({[(tert-butoxy)carbonyl]amino})hexanoic acid; N-cyclohexylcyclohexanamine
Molecular FormulaC28H53N3O6
InChI KeyHRLHJTYAMCGERD-VAMKTSTMNA-N
SMILESC1CCC(CC1)NC1CCCCC1.CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Proton NMR>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only