Thermo Scientific Chemicals

cis-2-(Boc-amino)cyclopropanemethanol, 97%, Thermo Scientific Chemicals

Catalog number: H62795.MD
250 mg, Each
Thermo Scientific Chemicals

cis-2-(Boc-amino)cyclopropanemethanol, 97%, Thermo Scientific Chemicals

Catalog number: H62795.MD
250 mg, Each
Quantity
Catalog number: H62795.MD
also known as H62795-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
170299-61-3
IUPAC Name
tert-butyl N-[2-(hydroxymethyl)cyclopropyl]carbamate
Molecular Formula
C9H17NO3
InChI Key
OCKKMJSVLVALMF-UHFFFAOYNA-N
SMILES
CC(C)(C)OC(=O)NC1CC1CO
Form
Solid
Carbon-13 NMR
Conforms to structure
Proton NMR
Conforms to structure
Appearance (Color)
Pale brown
Assay (GC)
>97%

Description

cis-2-(Boc-amino)cyclopropanemethanol is employed as a intermediate for chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-2-(Boc-amino)cyclopropanemethanol is employed as a intermediate for chemical research.

Notes
Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text