Thermo Scientific Chemicals

Potassium trimethylacetate, 95%, Thermo Scientific Chemicals

Catalog number: H62803.14
25 g, Each
Thermo Scientific Chemicals

Potassium trimethylacetate, 95%, Thermo Scientific Chemicals

Catalog number: H62803.14
25 g, Each
Quantity
Catalog number: H62803.14
also known as H62803-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
19455-23-3
IUPAC Name
potassium 2,2-dimethylpropanoate
Molecular Formula
C5H9KO2
InChI Key
WFMNHCSATCWAAQ-UHFFFAOYSA-M
SMILES
[K+].CC(C)(C)C([O-])=O
Assay (unspecified)
≥94.0%
Form
Powder or crystals or crystalline powder

Description

Potassium trimethylacetate is used as a intermediate for chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium trimethylacetate is used as a intermediate for chemical research.

Notes
Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text