Ethyl 1-(hydroxymethyl)cyclopentanecarboxylate, 97%, Thermo Scientific Chemicals
Ethyl 1-(hydroxymethyl)cyclopentanecarboxylate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 1-(hydroxymethyl)cyclopentanecarboxylate, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 g
Catalog number H62804.03
also known as H62804-03
Price (USD)/ Each
325.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
325.00
Add to cart
Ethyl 1-(hydroxymethyl)cyclopentanecarboxylate, 97%, Thermo Scientific Chemicals
Catalog numberH62804.03
Price (USD)/ Each
325.00
-
Add to cart
Specifications
Chemical Name or MaterialEthyl 1-(hydroxymethyl)cyclopentanecarboxylate
CAS1075-82-7
Recommended StorageAmbient temperatures
TSCANo
Molecular FormulaC9H16O3
View more
Ethyl 1-(hydroxymethyl)cyclopentanecarboxylate is employed as a intermediate for chemical research and synthetic building block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 1-(hydroxymethyl)cyclopentanecarboxylate is employed as a intermediate for chemical research and synthetic building block.

Notes
Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only