Thermo Scientific Chemicals

N-Fmoc-N-methylglycine, 98%, Thermo Scientific Chemicals

Catalog number: H62806.06
5 g, Each
Thermo Scientific Chemicals

N-Fmoc-N-methylglycine, 98%, Thermo Scientific Chemicals

Catalog number: H62806.06
5 g, Each
Quantity
Catalog number: H62806.06
also known as H62806-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
77128-70-2
IUPAC Name
2-({[(9H-fluoren-9-yl)methoxy]carbonyl}(methyl)amino)acetic acid hydrate
Molecular Formula
C18H19NO5
InChI Key
CUJSWOOWOONPRH-UHFFFAOYSA-N
SMILES
O.CN(CC(O)=O)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Assay (HPLC)
>97.5%

Description

N-Fmoc-N-methylglycine is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-N-methylglycine is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in DMF.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents, heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text