Ndelta-Boc-Nalpha-Fmoc-L-ornithine, 96%, Thermo Scientific Chemicals
Ndelta-Boc-Nalpha-Fmoc-L-ornithine, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ndelta-Boc-Nalpha-Fmoc-L-ornithine, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number H62814.14
also known as H62814-14
Price (USD)/ Each
404.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
404.00
Add to cart
Ndelta-Boc-Nalpha-Fmoc-L-ornithine, 96%, Thermo Scientific Chemicals
Catalog numberH62814.14
Price (USD)/ Each
404.00
-
Add to cart
Chemical Identifiers
CAS109425-55-0
IUPAC Name5-{[(tert-butoxy)carbonyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)pentanoic acid
Molecular FormulaC25H30N2O6
InChI KeyJOOIZTMAHNLNHE-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)NCCCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>95.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only