Thermo Scientific Chemicals

(R)-4-Benzyloxycarbonylamino-2-(Boc-amino)butyric acid dicyclohexylammonium salt, 98%, Thermo Scientific Chemicals

Catalog number: H62817.06
5 g, Each
Thermo Scientific Chemicals

(R)-4-Benzyloxycarbonylamino-2-(Boc-amino)butyric acid dicyclohexylammonium salt, 98%, Thermo Scientific Chemicals

Catalog number: H62817.06
5 g, Each
Quantity
Catalog number: H62817.06
also known as H62817-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
101854-42-6
IUPAC Name
4-{[(benzyloxy)carbonyl]amino}-2-{[(tert-butoxy)carbonyl]amino}butanoic acid; N-cyclohexylcyclohexanamine
Molecular Formula
C29H47N3O6
InChI Key
CBSVEVKFQHTZSP-UHFFFAOYNA-N
SMILES
C1CCC(CC1)NC1CCCCC1.CC(C)(C)OC(=O)NC(CCNC(=O)OCC1=CC=CC=C1)C(O)=O
Assay (HPLC)
>97.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text