Thermo Scientific Chemicals

N-Benzyloxycarbonylglycine methyl ester, 98%, Thermo Scientific Chemicals

Catalog number: H62858.14
25 g, Each
Thermo Scientific Chemicals

N-Benzyloxycarbonylglycine methyl ester, 98%, Thermo Scientific Chemicals

Catalog number: H62858.14
25 g, Each
Quantity
Catalog number: H62858.14
also known as H62858-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1212-53-9
IUPAC Name
methyl 2-{[(benzyloxy)carbonyl]amino}acetate
Molecular Formula
C11H13NO4
InChI Key
DZYBBBYFLOPVOL-UHFFFAOYSA-N
SMILES
COC(=O)CNC(=O)OCC1=CC=CC=C1
Appearance (Color)
Yellow to pale yellow
Form
Low melting solid
Assay (HPLC)
>97.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text