N-Boc-glycylglycine, 97%, Thermo Scientific Chemicals
N-Boc-glycylglycine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-glycylglycine, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Catalog number H62889.06
also known as H62889-06
Price (USD)/ Each
132.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
132.00
Add to cart
N-Boc-glycylglycine, 97%, Thermo Scientific Chemicals
Catalog numberH62889.06
Price (USD)/ Each
132.00
-
Add to cart
Chemical Identifiers
CAS31972-52-8
IUPAC Name2-(2-{[(tert-butoxy)carbonyl]amino}acetamido)acetic acid
Molecular FormulaC9H16N2O5
InChI KeyHWBAHOVOSOAFLE-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)NCC(=O)NCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only