2-Carbamoyl-3-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals
2-Carbamoyl-3-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Carbamoyl-3-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H62904.MD
also known as H62904-MD
Price (USD)/ Each
194.00
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
194.00
Add to cart
2-Carbamoyl-3-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals
Catalog numberH62904.MD
Price (USD)/ Each
194.00
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or Material2-Carbamoyl-3-fluorobenzeneboronic acid pinacol ester
Recommended StorageAmbient temperatures
TSCANo
Quantity250 mg
Synonym2-Carbamoyl-3-fluorophenylboronic acid pinacol ester
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only