DL-Glutamine, 98%, Thermo Scientific Chemicals
DL-Glutamine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

DL-Glutamine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H62915.03
also known as H62915-03
Price (USD)/ Each
41.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
48.70 
Save 7.05 (14%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
41.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
48.70 
Save 7.05 (14%)
Add to cart
DL-Glutamine, 98%, Thermo Scientific Chemicals
Catalog numberH62915.03
Price (USD)/ Each
41.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
48.70 
Save 7.05 (14%)
-
Add to cart
Chemical Identifiers
CAS6899-04-3
IUPAC Name2-amino-4-carbamoylbutanoic acid
Molecular FormulaC5H10N2O3
InChI KeyZDXPYRJPNDTMRX-UHFFFAOYNA-N
SMILESNC(CCC(N)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay from Suppliers CofA≥97.5%
FormPowder or crystals or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only