cis-4-(Boc-amino)cyclohexylamine, 97%, Thermo Scientific Chemicals
cis-4-(Boc-amino)cyclohexylamine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

cis-4-(Boc-amino)cyclohexylamine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H62925.06
also known as H62925-06
Price (USD)/ Each
231.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
231.00
Add to cart
cis-4-(Boc-amino)cyclohexylamine, 97%, Thermo Scientific Chemicals
Catalog numberH62925.06
Price (USD)/ Each
231.00
-
Add to cart
Specifications
Chemical Name or Materialcis-4-(Boc-amino)cyclohexylamine
CAS247570-24-7
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
cis-4-(Boc-amino)cyclohexylamine can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-4-(Boc-amino)cyclohexylamine can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in methanol, ethyl acetate and Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only