Thermo Scientific Chemicals

cis-4-(Boc-amino)cyclohexylamine, 97%, Thermo Scientific Chemicals

Catalog number: H62925.14
25 g, Each
Thermo Scientific Chemicals

cis-4-(Boc-amino)cyclohexylamine, 97%, Thermo Scientific Chemicals

Catalog number: H62925.14
25 g, Each
Quantity
Catalog number: H62925.14
also known as H62925-14
Price (USD)
Quantity
-
Have Questions?

Description

cis-4-(Boc-amino)cyclohexylamine can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-4-(Boc-amino)cyclohexylamine can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in methanol, ethyl acetate and Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text