N-Fmoc-L-glutamic acid, 95%, Thermo Scientific Chemicals
N-Fmoc-L-glutamic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Fmoc-L-glutamic acid, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number H62936.14
also known as H62936-14
Price (USD)/ Each
145.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
145.00
Add to cart
N-Fmoc-L-glutamic acid, 95%, Thermo Scientific Chemicals
Catalog numberH62936.14
Price (USD)/ Each
145.00
-
Add to cart
Chemical Identifiers
CAS121343-82-6
IUPAC Name2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)pentanedioic acid
Molecular FormulaC20H19NO6
InChI KeyQEPWHIXHJNNGLU-UHFFFAOYNA-N
SMILESOC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (HPLC)>94.0%
FormPowder
N-Fmoc-L-glutamic acid nonessential amino acid that is important to the mammalian central nervous system. It is also a neurotransmitter for cone photoreceptors in the human brain and is used as a treatment for patients who have liver disease accompanied by encephalopathy (a condition termed ‘hepatic coma’).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Tomsho JW et al. Synthesis of (6R)- and (6S)-5,10-dideazatetrahydrofolate oligo-?-glutamates: Kinetics of multiple glutamate ligations catalyzed by folylpoly-?-glutamate synthetase. Organic & biomolecular chemistry. 2005, 3 (18), 3388-3398.
  2. Yuko Kamikawa; Masayuki Nishii; and Takashi Kato Prof. Self-Assembly of Folic Acid Derivatives: Induction of Supramolecular Chirality by Hierarchical Chiral Structures. Chemistry - A European Journal. 2004, 10 (23), 5942-5951.