N-Boc-4-cyano-D-phenylalanine, 98%, Thermo Scientific Chemicals
N-Boc-4-cyano-D-phenylalanine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-4-cyano-D-phenylalanine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H62947.03
also known as H62947-03
Price (USD)/ Each
133.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
133.00
Add to cart
N-Boc-4-cyano-D-phenylalanine, 98%, Thermo Scientific Chemicals
Catalog numberH62947.03
Price (USD)/ Each
133.00
-
Add to cart
Chemical Identifiers
CAS146727-62-0
IUPAC Name(2R)-2-{[(tert-butoxy)carbonyl]amino}-3-(4-cyanophenyl)propanoate
Molecular FormulaC15H17N2O4
InChI KeyRMBLTLXJGNILPG-GFCCVEGCSA-M
SMILESCC(C)(C)OC(=O)N[C@H](CC1=CC=C(C=C1)C#N)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
N-Boc-4-cyano-D-phenylalanine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-4-cyano-D-phenylalanine is used as pharmaceutical intermediate.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only