trans-4-(Benzyloxycarbonylamino)cyclohexanecarboxaldehyde, 97%, Thermo Scientific Chemicals
trans-4-(Benzyloxycarbonylamino)cyclohexanecarboxaldehyde, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

trans-4-(Benzyloxycarbonylamino)cyclohexanecarboxaldehyde, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H62957.06
also known as H62957-06
Price (USD)/ Each
1,069.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
1,069.00
Add to cart
trans-4-(Benzyloxycarbonylamino)cyclohexanecarboxaldehyde, 97%, Thermo Scientific Chemicals
Catalog numberH62957.06
Price (USD)/ Each
1,069.00
-
Add to cart
Specifications
Chemical Name or Materialtrans-4-(Benzyloxycarbonylamino)cyclohexanecarboxaldehyde
CAS412357-50-7
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only