Thermo Scientific Chemicals

Nepsilon-Boc-L-lysine tert-butyl ester hydrochloride, 95%, Thermo Scientific Chemicals

Catalog number: H62993.06
5 g, Each
Thermo Scientific Chemicals

Nepsilon-Boc-L-lysine tert-butyl ester hydrochloride, 95%, Thermo Scientific Chemicals

Catalog number: H62993.06
5 g, Each
Quantity
Catalog number: H62993.06
also known as H62993-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13288-57-8
IUPAC Name
tert-butyl (2S)-2-amino-6-{[(tert-butoxy)carbonyl]amino}hexanoate hydrochloride
Molecular Formula
C15H31ClN2O4
InChI Key
TZBPQINFXPIRBX-VAMKTSTMNA-N
SMILES
Cl.CC(C)(C)OC(=O)NCCCC[C@H](N)C(=O)OC(C)(C)C
Assay (HPLC)
>94.0%

Description

Nepsilon-Boc-L-lysine tert-butyl ester hydrochloride is an pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nepsilon-Boc-L-lysine tert-butyl ester hydrochloride is an pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text