DL-tert-Leucine, 95%, Thermo Scientific Chemicals
DL-tert-Leucine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

DL-tert-Leucine, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H63007.03
also known as H63007-03
Price (USD)/ Each
50.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
50.00
Add to cart
DL-tert-Leucine, 95%, Thermo Scientific Chemicals
Catalog numberH63007.03
Price (USD)/ Each
50.00
-
Add to cart
Chemical Identifiers
CAS33105-81-6
IUPAC Name2-amino-3,3-dimethylbutanoic acid
Molecular FormulaC6H13NO2
InChI KeyNPDBDJFLKKQMCM-UHFFFAOYSA-N
SMILESCC(C)(C)C(N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only