Nepsilon-2,4-Dinitrophenyl-Nalpha-Fmoc-L-lysine, 98%, Thermo Scientific Chemicals
Nepsilon-2,4-Dinitrophenyl-Nalpha-Fmoc-L-lysine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nepsilon-2,4-Dinitrophenyl-Nalpha-Fmoc-L-lysine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H63050.03
also known as H63050-03
Price (USD)/ Each
144.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
144.00
Add to cart
Nepsilon-2,4-Dinitrophenyl-Nalpha-Fmoc-L-lysine, 98%, Thermo Scientific Chemicals
Catalog numberH63050.03
Price (USD)/ Each
144.00
-
Add to cart
Chemical Identifiers
CAS148083-64-1
IUPAC Name(2S)-6-[(2,4-dinitrophenyl)amino]-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
Molecular FormulaC27H26N4O8
InChI KeyOENCMORJQAUAAJ-FQKVKQEKNA-N
SMILESOC(=O)[C@H](CCCCNC1=C(C=C(C=C1)[N+]([O-])=O)[N+]([O-])=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only