2-(4-Chlorophenyl)ethylamine, 97%, Thermo Scientific Chemicals
2-(4-Chlorophenyl)ethylamine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-(4-Chlorophenyl)ethylamine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number H63081.14
also known as H63081-14
Price (USD)/ Each
125.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
125.00
Add to cart
2-(4-Chlorophenyl)ethylamine, 97%, Thermo Scientific Chemicals
Catalog numberH63081.14
Price (USD)/ Each
125.00
-
Add to cart
Specifications
Chemical Name or Material2-(4-Chlorophenyl)ethylamine
CAS156-41-2
Health Hazard 1H314-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only