O-Methyl-L-tyrosine, 98%, Thermo Scientific Chemicals
O-Methyl-L-tyrosine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

O-Methyl-L-tyrosine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H63096.03
also known as H63096-03
Price (USD)/ Each
32.90
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
32.90
Add to cart
O-Methyl-L-tyrosine, 98%, Thermo Scientific Chemicals
Catalog numberH63096.03
Price (USD)/ Each
32.90
-
Add to cart
Chemical Identifiers
CAS6230-11-1
IUPAC Name(2S)-2-amino-3-(4-methoxyphenyl)propanoic acid
Molecular FormulaC10H13NO3
InChI KeyGEYBMYRBIABFTA-IMWMWJONNA-N
SMILESCOC1=CC=C(C[C@H](N)C(O)=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
O-Methyl-L-tyrosine, is used as a fine chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-Methyl-L-tyrosine, is used as a fine chemical intermediate.

Solubility
Very soluble in water.

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Y Zhang.; L Wang.; PG Schultz.; IA Wilson. Crystal structures of apo wild-type M. jannaschii tyrosyl-tRNA synthetase (TyrRS) and an engineered TyrRS specific for O-methyl-L-tyrosine. Protein science.200514 (5), 1340-1345.
  2. L Wang.; A Brock.; B Herberich.; PG Schultz. Expanding the genetic code of Escherichia coli. Science.2001292 (5516), 498-500 .