Thermo Scientific Chemicals

O-Methyl-L-tyrosine, 98%, Thermo Scientific Chemicals

Catalog number: H63096.06
5 g, Each
Thermo Scientific Chemicals

O-Methyl-L-tyrosine, 98%, Thermo Scientific Chemicals

Catalog number: H63096.06
5 g, Each
Quantity
Catalog number: H63096.06
also known as H63096-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6230-11-1
IUPAC Name
(2S)-2-amino-3-(4-methoxyphenyl)propanoic acid
Molecular Formula
C10H13NO3
InChI Key
GEYBMYRBIABFTA-IMWMWJONNA-N
SMILES
COC1=CC=C(C[C@H](N)C(O)=O)C=C1
Assay (HPLC)
>97.5%

Description

O-Methyl-L-tyrosine, is used as a fine chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-Methyl-L-tyrosine, is used as a fine chemical intermediate.

Solubility
Very soluble in water.

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text