5-Methoxy-7-methylindole, 97%, Thermo Scientific Chemicals
5-Methoxy-7-methylindole, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Methoxy-7-methylindole, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H63123.06
also known as H63123-06
Price (USD)/ Each
895.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
895.00
Add to cart
5-Methoxy-7-methylindole, 97%, Thermo Scientific Chemicals
Catalog numberH63123.06
Price (USD)/ Each
895.00
-
Add to cart
Specifications
Chemical Name or Material5-Methoxy-7-methylindole
CAS61019-05-4
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
5-Methoxy-7-methylindole is employed as a intermediate for chemical research and pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methoxy-7-methylindole is employed as a intermediate for chemical research and pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Rüdiger Faust; Peter J. Garratt; Maria Angeles Trujillo Pérez; Vincent J.-D. Piccio; Christian Madsen; Ane Stenstrøm; Bente Frølund; Kathryn Davidson; Muy-Teck Teh; David Sugden. 7-Substituted-melatonin and 7-substituted-1-methylmelatonin analogues: Effect of substituents on potency and binding affinity. Bioorganic & Medicinal Chemistry. 2007, 15(13), 4543-4551.