6-(Trifluoromethyl)pyridine-2-carboxylic acid, 98%, Thermo Scientific Chemicals
6-(Trifluoromethyl)pyridine-2-carboxylic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-(Trifluoromethyl)pyridine-2-carboxylic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number H63138.03
also known as H63138-03
Price (USD)/ Each
119.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
119.00
Add to cart
6-(Trifluoromethyl)pyridine-2-carboxylic acid, 98%, Thermo Scientific Chemicals
Catalog numberH63138.03
Price (USD)/ Each
119.00
-
Add to cart
Chemical Identifiers
CAS131747-42-7
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only