Thermo Scientific Chemicals

6-(Fmoc-amino)hexanoic acid, 98%, Thermo Scientific Chemicals

Catalog number: H63148.03
1 g, Each
Thermo Scientific Chemicals

6-(Fmoc-amino)hexanoic acid, 98%, Thermo Scientific Chemicals

Catalog number: H63148.03
1 g, Each
Quantity
Catalog number: H63148.03
also known as H63148-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
88574-06-5
IUPAC Name
6-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
Molecular Formula
C21H23NO4
InChI Key
FPCPONSZWYDXRD-UHFFFAOYSA-N
SMILES
OC(=O)CCCCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Assay (HPLC)
>97.5%

Description

6-(Fmoc-amino)hexanoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-(Fmoc-amino)hexanoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in 1% acetic acid, water and 1mmol in 2mL DMF clearly soluble.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text