Thermo Scientific Chemicals

6-Amino-5-bromoquinoline, 97%, Thermo Scientific Chemicals

Catalog number: H63150.06
5 g, Each
Thermo Scientific Chemicals

6-Amino-5-bromoquinoline, 97%, Thermo Scientific Chemicals

Catalog number: H63150.06
5 g, Each
Quantity
Catalog number: H63150.06
also known as H63150-06
Price (USD)
Quantity
-
Have Questions?

Description

6-Amino-5-bromoquinoline, has a wide range of biological and pharmacological activities. It shows both electrophilic and nucleophilic substitution reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Amino-5-bromoquinoline, has a wide range of biological and pharmacological activities. It shows both electrophilic and nucleophilic substitution reactions.

Solubility
Slightly soluble in water. Soluble in alcohol, ether, and carbon disulfide.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text