Thermo Scientific Chemicals

Sarcosine tert-butyl ester hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: H63176.14
25 g, Each
Thermo Scientific Chemicals

Sarcosine tert-butyl ester hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: H63176.14
25 g, Each
Quantity
Catalog number: H63176.14
also known as H63176-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5616-81-9
IUPAC Name
tert-butyl 2-(methylamino)acetate hydrochloride
Molecular Formula
C7H16ClNO2
InChI Key
RNLQHMIDSCYLAK-UHFFFAOYSA-N
SMILES
Cl.CNCC(=O)OC(C)(C)C
Assay (HPLC)
>96.0%

Description

Sarcosine tert-butyl ester hydrochloride, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sarcosine tert-butyl ester hydrochloride, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in methanol (0.3 gram in 2 mL MeOH).

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text