Thermo Scientific Chemicals

3,4-Dimethylphenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: H63178.06
5 g, Each
Thermo Scientific Chemicals

3,4-Dimethylphenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: H63178.06
5 g, Each
Quantity
Catalog number: H63178.06
also known as H63178-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
17283-16-8
Assay (HPLC)
>97.5%

Description

3,4-Dimethylphenylacetic acid, is is used as an intermediate in organic synthesis. It is also used as a fine chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethylphenylacetic acid, is is used as an intermediate in organic synthesis. It is also used as a fine chemical intermediate.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text