D-Aspartic acid dimethyl ester hydrochloride, 98%, Thermo Scientific Chemicals
D-Aspartic acid dimethyl ester hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

D-Aspartic acid dimethyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H63197.06
also known as H63197-06
Price (USD)/ Each
148.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
148.00
Add to cart
D-Aspartic acid dimethyl ester hydrochloride, 98%, Thermo Scientific Chemicals
Catalog numberH63197.06
Price (USD)/ Each
148.00
-
Add to cart
Chemical Identifiers
CAS69630-50-8
IUPAC Name1,4-dimethyl 2-aminobutanedioate hydrochloride
Molecular FormulaC6H12ClNO4
InChI KeyPNLXWGDXZOYUKB-UHFFFAOYNA-N
SMILESCl.COC(=O)CC(N)C(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only