Thermo Scientific Chemicals

N-Boc-guanidine, 95%, Thermo Scientific Chemicals

Catalog number: H63206.06
5 g, Each
Thermo Scientific Chemicals

N-Boc-guanidine, 95%, Thermo Scientific Chemicals

Catalog number: H63206.06
5 g, Each
Quantity
Catalog number: H63206.06
also known as H63206-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
219511-71-4
IUPAC Name
tert-butyl N-(diaminomethylidene)carbamate
Molecular Formula
C6H13N3O2
InChI Key
UMOZLQVSOVNSCA-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)N=C(N)N
Assay (unspecified)
≥94.0%
Form
Powder
Appearance (Color)
White to pale yellow

Description

N-Boc-guanidine, is used as used as guanidinylating agent and for synthesis of 2-aminoimidazoles. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-guanidine, is used as used as guanidinylating agent and for synthesis of 2-aminoimidazoles. It is also used as a pharmaceutical intermediate.

Solubility
Soluble in most alcohols, esters (e.g., ethyl acetate), ketones (e.g., acetone), acetonitrile, and dichloromethane. Not soluble in diethyl ether and hexane.

Notes
Moisture, heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text