Ethyl 2-methylquinoline-6-carboxylate, 97%, Thermo Scientific Chemicals
Ethyl 2-methylquinoline-6-carboxylate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 2-methylquinoline-6-carboxylate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H63213.03
also known as H63213-03
Price (USD)/ Each
59.60
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
59.60
Add to cart
Ethyl 2-methylquinoline-6-carboxylate, 97%, Thermo Scientific Chemicals
Catalog numberH63213.03
Price (USD)/ Each
59.60
-
Add to cart
Chemical Identifiers
CAS855763-77-8
IUPAC Nameethyl 2-methylquinoline-6-carboxylate
Molecular FormulaC13H13NO2
InChI KeyWLKKUWXLOSMSMH-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CC=C2N=C(C)C=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only