4-Bromomethyl-3-fluorobenzonitrile, 95%, Thermo Scientific Chemicals
4-Bromomethyl-3-fluorobenzonitrile, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Bromomethyl-3-fluorobenzonitrile, 95%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Catalog number H63245.06
also known as H63245-06
Price (USD)/ Each
137.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
137.00
Add to cart
4-Bromomethyl-3-fluorobenzonitrile, 95%, Thermo Scientific Chemicals
Catalog numberH63245.06
Price (USD)/ Each
137.00
-
Add to cart
Chemical Identifiers
CAS105942-09-4
IUPAC Name4-(bromomethyl)-3-fluorobenzonitrile
Molecular FormulaC8H5BrFN
InChI KeyZESZAIOGACKOMB-UHFFFAOYSA-N
SMILESFC1=C(CBr)C=CC(=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Proton NMR>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only