Thermo Scientific Chemicals

N-Benzyloxycarbonyl-L-valine N-succinimidyl ester, 98%, Thermo Scientific Chemicals

Catalog number: H63253.14
25 g, Each
Thermo Scientific Chemicals

N-Benzyloxycarbonyl-L-valine N-succinimidyl ester, 98%, Thermo Scientific Chemicals

Catalog number: H63253.14
25 g, Each
Quantity
Catalog number: H63253.14
also known as H63253-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3496-11-5
IUPAC Name
2,5-dioxopyrrolidin-1-yl (2S)-2-{[(benzyloxy)carbonyl]amino}-3-methylbutanoate
Molecular Formula
C17H20N2O6
InChI Key
MFAOBGXYLNLLJE-HNNXBMFYSA-N
SMILES
CC(C)[C@H](NC(=O)OCC1=CC=CC=C1)C(=O)ON1C(=O)CCC1=O
Assay (HPLC)
>97.5%

Description

N-Benzyloxycarbonyl-L-valine N-succinimidyl ester, is an amino acid which is widely used in pharmaceutical and food industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzyloxycarbonyl-L-valine N-succinimidyl ester, is an amino acid which is widely used in pharmaceutical and food industry.

Solubility
Soluble in 1mmol in 2mL DCM.

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text