Methyl 4-fluoro-2-methylbenzoate, 98%, Thermo Scientific Chemicals
Methyl 4-fluoro-2-methylbenzoate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 4-fluoro-2-methylbenzoate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number H63268.06
also known as H63268-06
Price (USD)/ Each
45.80
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
45.80
Add to cart
Methyl 4-fluoro-2-methylbenzoate, 98%, Thermo Scientific Chemicals
Catalog numberH63268.06
Price (USD)/ Each
45.80
-
Add to cart
Chemical Identifiers
CAS174403-69-1
IUPAC Namemethyl 4-fluoro-2-methylbenzoate
Molecular FormulaC9H9FO2
InChI KeyZQFCTCYDRQFPBU-UHFFFAOYSA-N
SMILESCOC(=O)C1=C(C)C=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only