Thermo Scientific Chemicals

(S)-4-(1-Aminoethyl)phenol, 97%, Thermo Scientific Chemicals

Catalog number: H63269.06
5 g, Each
Thermo Scientific Chemicals

(S)-4-(1-Aminoethyl)phenol, 97%, Thermo Scientific Chemicals

Catalog number: H63269.06
5 g, Each
Quantity
Catalog number: H63269.06
also known as H63269-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
221670-72-0
Assay from Suppliers CofA
≥96.0% (Chiral HPLC)
Appearance (Color)
White to cream or gray
Form
Crystals or powder or crystalline powder
Proton NMR
Conforms to structure

Description

(S)-4-(1-Aminoethyl)phenol is used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-4-(1-Aminoethyl)phenol is used as an intermediate in organic synthesis.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text