Chemical Identifiers
CAS76265-70-8
IUPAC Name(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-methanesulfinylbutanoic acid
Molecular FormulaC20H21NO5S
InChI KeyCEHRSUBRZOGRSW-HSYKDVHTSA-N
SMILESCS(=O)CC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
View more