Thermo Scientific Chemicals

L-Glutamic acid 1-methyl ester, 98%, Thermo Scientific Chemicals

Catalog number: H63279.03
1 g, Each
Thermo Scientific Chemicals

L-Glutamic acid 1-methyl ester, 98%, Thermo Scientific Chemicals

Catalog number: H63279.03
1 g, Each
Quantity
Catalog number: H63279.03
also known as H63279-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6384-08-3
IUPAC Name
4-amino-5-methoxy-5-oxopentanoic acid
Molecular Formula
C6H11NO4
InChI Key
SEWIYICDCVPBEW-UHFFFAOYNA-N
SMILES
COC(=O)C(N)CCC(O)=O
Assay (HPLC)
>97.5%

Description

L-Glutamic acid 1-methyl ester nonessential amino acid that is important to the mammalian central nervous system. It is also a neurotransmitter for cone photoreceptors in the human brain and is used as a treatment for patients who have liver disease accompanied by encephalopathy (a condition termed ‘hepatic coma’).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text