Thermo Scientific Chemicals

D-Glutamic acid 1-tert-butyl ester, 98%, Thermo Scientific Chemicals

Catalog number: H63318.06
5 g, Each
Thermo Scientific Chemicals

D-Glutamic acid 1-tert-butyl ester, 98%, Thermo Scientific Chemicals

Catalog number: H63318.06
5 g, Each
Quantity
Catalog number: H63318.06
also known as H63318-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
25456-76-2
IUPAC Name
4-amino-5-(tert-butoxy)-5-oxopentanoic acid
Molecular Formula
C9H17NO4
InChI Key
QVAQMUAKTNUNLN-UHFFFAOYNA-N
SMILES
CC(C)(C)OC(=O)C(N)CCC(O)=O
Assay (HPLC)
>97.5%

Description

D-Glutamic acid 1-tert-butyl ester, is an aminoacid which is widely used in pharmaceutical and food industry. It is also used as a pharmaceutical intermediate. It is also an important organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Glutamic acid 1-tert-butyl ester, is an aminoacid which is widely used in pharmaceutical and food industry. It is also used as a pharmaceutical intermediate. It is also an important organic intermediate.

Solubility
Slightly soluble in water.

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text